Learn More
Aviva Systems Biology Corp CFDP1 Antibody - middle region (ARP52145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CFDP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CFDP1. Alternate names: BCNT, CP27, SWC5, Yeti, p97, CENP-29, BUCENTAURApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-649