Learn More
Enzo Life Sciences Complement factor H (beta1H-globulin) monoclonal antibody (C18/3) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDGAU018031
Encompass_Preferred
Clone: C18/3. Isotype: IgG1κ. Immunogen: Human complement factor H. Species reactivity: Human. Applications: ELISA, WB. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2462