Learn More
Aviva Systems Biology Corp CFHR1 Antibody - N-terminal region (ARP62261_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62261_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CFHR1. It was validated on Western Blot. . Alternate names: CFHL, CFHL1, CFHL1P, CFHR1P, FHR1, H36-1, H36-2, HFL1, HFL2, MGC104329ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-287-634