Learn More
Apexbio Technology LLC CFI-400945 1338806-73-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064300
Encompass_Preferred
CFI-400945 (CAS 1338806-73-7) is an orally bioavailable inhibitor specifically targeting polo-like kinase 4 (PLK4) a member of the polo-like kinase family involved in centriole duplication and cell cycle control By competitively inhibiting ATP binding to PLK4 CFI-400945 potently suppresses PLK4 catalytic activity (IC50 2 8 nM) and blocks PLK4 autophosphorylation in cellular assays (EC50 12 3 nM) The compound exhibits high selectivity against other polo-like kinases and demonstrates antiproliferative activity in various cancer cell lines Thus CFI-400945 is applied in cancer biology research particularly for studying centrosome regulation and tumor growth inhibition
Catalog No. 50-000-64300 Supplier Apexbio Technology LLC Supplier No. C58135.1
CFI-400945 (CAS 1338806-73-7) is an orally bioavailable inhibitor specifically targeting polo-like kinase 4 (PLK4) a member of the polo-like kinase family involved in centriole duplication and cell cycle control By competitively inhibiting ATP binding to PLK4 CFI-400945 potently suppresses PLK4 catalytic activity (IC50 2 8 nM) and blocks PLK4 autophosphorylation in cellular assays (EC50 12 3 nM) The compound exhibits high selectivity against other polo-like kinases and demonstrates antiproliferative activity in various cancer cell lines Thus CFI-400945 is applied in cancer biology research particularly for studying centrosome regulation and tumor growth inhibition