Learn More
Medchemexpress LLC CFL-120 | 18711-15-4 | ≥98.0% | 216.02 g/mol | C8H3Cl2NO2 | 25 G
SDP
Catalog No. 5000208907
Encompass_Preferred
CFL-120 is an allele-specific covalent small-molecule inhibitor that selectively targets the KRas G12C mutant. It shows antiproliferative and anticancer activity in preclinical models and is supplied for research use only.
  • Selective covalent inhibitor of KRas G12C.
  • Demonstrates antiproliferative activity in cell-based assays.
  • Useful for preclinical studies of KRas G12C-driven lung cancer.
  • High chemical purity (≥98.0%).
  • Available in multiple package sizes, including 25 g.
Catalog No. 50-002-08907 Supplier Medchemexpress LLC Supplier No. 25GHYW041315
CFL-120 is an allele-specific covalent small-molecule inhibitor that selectively targets the KRas G12C mutant. It shows antiproliferative and anticancer activity in preclinical models and is supplied for research use only.
  • Selective covalent inhibitor of KRas G12C.
  • Demonstrates antiproliferative activity in cell-based assays.
  • Useful for preclinical studies of KRas G12C-driven lung cancer.
  • High chemical purity (≥98.0%).
  • Available in multiple package sizes, including 25 g.