Learn More
Grainger CFL Ballast Instant Start 40W 2 Bulbs
Catalog No. 502767918
Encompass_Preferred
The Lumapro CFL Ballast Compact Fluorescent is a 16 watt, 120 volt electronic ballast that operates one 13-42 watt 4-pin CFL lamp. It has an input frequency of 60 Hz and features instant start technology for immediate light output. This electronic ballast has a thermally protected class P insulation system and universal voltage capability. It measures 5 inches long by 1.25 inches wide by 1.25 inches high. The ballast housing is made of UL94V-0 flame retardant plastic. This product is ETL/cETL listed and has a 5 year warranty.
Catalog No. 50-276-7918 Supplier Grainger Supplier No. 16X955
The Lumapro CFL Ballast Compact Fluorescent is a 16 watt, 120 volt electronic ballast that operates one 13-42 watt 4-pin CFL lamp. It has an input frequency of 60 Hz and features instant start technology for immediate light output. This electronic ballast has a thermally protected class P insulation system and universal voltage capability. It measures 5 inches long by 1.25 inches wide by 1.25 inches high. The ballast housing is made of UL94V-0 flame retardant plastic. This product is ETL/cETL listed and has a 5 year warranty.