Learn More
Grainger CFL Ballast Programmed Start 18W 2 Bulbs
Catalog No. 502766988
Encompass_Preferred
The LUMAPRO CFL Ballast Compact Fluorescent is a 16 watt 120 volt electronic ballast that operates one 13-42 watt 4-pin CFL lamp. It has an input voltage range of 108-132V, input frequency of 50/60 Hz, and power factor greater than 0.9. This electronic ballast features anti-striation circuitry, end of lamp life shutdown, and total harmonic distortion less than 20%. It measures 5.25 inches long x 1.25 inches wide x 1.06 inches high and weighs 0.35 lbs. This ballast is cULus listed and comes with a 5 year warranty from Lumapro.
Catalog No. 50-276-6988 Supplier Grainger Supplier No. 16X959
The LUMAPRO CFL Ballast Compact Fluorescent is a 16 watt 120 volt electronic ballast that operates one 13-42 watt 4-pin CFL lamp. It has an input voltage range of 108-132V, input frequency of 50/60 Hz, and power factor greater than 0.9. This electronic ballast features anti-striation circuitry, end of lamp life shutdown, and total harmonic distortion less than 20%. It measures 5.25 inches long x 1.25 inches wide x 1.06 inches high and weighs 0.35 lbs. This ballast is cULus listed and comes with a 5 year warranty from Lumapro.