Learn More
Grainger CFL Ballast Programmed Start 26W 2 Bulbs
Catalog No. 502768226
Encompass_Preferred
The CFL Ballast Compact Fluorescent 120 to 277V AC 1 to 2 Bulbs 26 W Bulb Max is a ballast for compact fluorescent lamps. It is designed for 120 to 277 volt AC operation and can power 1 or 2 compact fluorescent bulbs up to 26 watts each. This electronic ballast provides reliable starting and operation of compact fluorescent lamps. It's engineered for long life and energy efficiency. This ballast measures 5-1/4" x 2-1/4" x 1-1/4" and comes with mounting hardware. It's CSA certified and ideal for energy efficient fluorescent lighting in commercial, industrial and residential applications.
Catalog No. 50-276-8226 Supplier Grainger Supplier No. 16X960
The CFL Ballast Compact Fluorescent 120 to 277V AC 1 to 2 Bulbs 26 W Bulb Max is a ballast for compact fluorescent lamps. It is designed for 120 to 277 volt AC operation and can power 1 or 2 compact fluorescent bulbs up to 26 watts each. This electronic ballast provides reliable starting and operation of compact fluorescent lamps. It's engineered for long life and energy efficiency. This ballast measures 5-1/4" x 2-1/4" x 1-1/4" and comes with mounting hardware. It's CSA certified and ideal for energy efficient fluorescent lighting in commercial, industrial and residential applications.