Learn More
Grainger CFL Ballast Programmed Start 40W 2 Bulbs
Catalog No. 502766991
Encompass_Preferred
The Lumapro CFL Ballast Compact Fluorescent is a 16 watt 120 volt electronic ballast that operates one 13-42 watt 4-pin CFL lamp. It has a thermoplastic housing, measures 5.5 inches long x 1.75 inches wide x 1.25 inches high, and weighs 5.8 ounces. This ballast has a 0.99 ballast factor, class P thermally protected, type A sound rating, and complies with FCC part 18. It is cULus listed and has a 5 year warranty. This ballast is ideal for operating energy saving CFL lamps.
Catalog No. 50-276-6991 Supplier Grainger Supplier No. 16X956
The Lumapro CFL Ballast Compact Fluorescent is a 16 watt 120 volt electronic ballast that operates one 13-42 watt 4-pin CFL lamp. It has a thermoplastic housing, measures 5.5 inches long x 1.75 inches wide x 1.25 inches high, and weighs 5.8 ounces. This ballast has a 0.99 ballast factor, class P thermally protected, type A sound rating, and complies with FCC part 18. It is cULus listed and has a 5 year warranty. This ballast is ideal for operating energy saving CFL lamps.