Learn More
Grainger CFL Ballast Programmed Start 42W 2 Bulbs
Catalog No. 502766865
Encompass_Preferred
The Grainger CFL Ballast Programmed Start 42W 2 Bulbs is an electronic ballast designed for use with compact fluorescent lamps. It operates 2 lamps at 42 watts total and provides programmed start operation, which preheats the lamp cathodes before ignition to extend lamp life. This 120V ballast has an input frequency of 60 Hz and is compatible with F42T12/ES lamps. It measures 11-1/4" x 2-1/2" x 1-1/4" and is cULus listed. The ballast comes with installation instructions and is backed by a 5 year warranty from Grainger.
Catalog No. 50-276-6865 Supplier Grainger Supplier No. 16X961
The Grainger CFL Ballast Programmed Start 42W 2 Bulbs is an electronic ballast designed for use with compact fluorescent lamps. It operates 2 lamps at 42 watts total and provides programmed start operation, which preheats the lamp cathodes before ignition to extend lamp life. This 120V ballast has an input frequency of 60 Hz and is compatible with F42T12/ES lamps. It measures 11-1/4" x 2-1/2" x 1-1/4" and is cULus listed. The ballast comes with installation instructions and is backed by a 5 year warranty from Grainger.