Learn More
Grainger CFL Ballast Rapid Start 13W 2 Bulbs
Catalog No. 502766800
Encompass_Preferred
The Lumapro CFL Ballast Compact Fluorescent is a 16W 120V electronic ballast that operates one 13W-42W 4-pin CFL lamp. It has a thermoplastic housing, measures 5-1/4" length x 2-1/4" width x 1-1/8" height, and weighs 0.55 lbs. This ballast has a 0.99 ballast factor, >0.95 power factor, <20% total harmonic distortion, and complies with FCC rules. It can be used with dimmable CFLs and has a 3-year warranty when used with Lumapro lamps. This electronic ballast is designed for indoor use.
Catalog No. 50-276-6800 Supplier Grainger Supplier No. 16X952
The Lumapro CFL Ballast Compact Fluorescent is a 16W 120V electronic ballast that operates one 13W-42W 4-pin CFL lamp. It has a thermoplastic housing, measures 5-1/4" length x 2-1/4" width x 1-1/8" height, and weighs 0.55 lbs. This ballast has a 0.99 ballast factor, >0.95 power factor, <20% total harmonic distortion, and complies with FCC rules. It can be used with dimmable CFLs and has a 3-year warranty when used with Lumapro lamps. This electronic ballast is designed for indoor use.