Learn More
Grainger CFL Ballast Rapid Start 18W 2 Bulbs
Catalog No. 502766801
Encompass_Preferred
The Grainger CFL Ballast Rapid Start 18W 2 Bulbs is an electronic ballast designed for use with compact fluorescent lamps. It operates two 18W T8 twin tube lamps and provides instant start operation. This ballast has a Class P thermally protected case, offers quiet operation and high frequency operation at 20kHz. It is UL and cUL listed and has a 5 year warranty. The input voltage is 120V at 60Hz and the ballast factor is 1.0. This 18W rapid start ballast is ideal for energy efficient fluorescent lighting applications.
Catalog No. 50-276-6801 Supplier Grainger Supplier No. 16X953
The Grainger CFL Ballast Rapid Start 18W 2 Bulbs is an electronic ballast designed for use with compact fluorescent lamps. It operates two 18W T8 twin tube lamps and provides instant start operation. This ballast has a Class P thermally protected case, offers quiet operation and high frequency operation at 20kHz. It is UL and cUL listed and has a 5 year warranty. The input voltage is 120V at 60Hz and the ballast factor is 1.0. This 18W rapid start ballast is ideal for energy efficient fluorescent lighting applications.