Learn More
Aviva Systems Biology Corp CFLAR Antibody - C-terminal region (AVARP02011_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02011_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CFLAR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CFLAR. Alternate names: CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLIP, I-FLICE, MRIT, USURPIN, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS, FLAME1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62475