Learn More
Electron Microscopy Sciences C-Flat™ Holey Carbon Grid CF-2/4-4C-Thick, 400 mesh Cu
SDP
Catalog No. 501926559
Encompass_Preferred
C-flat is an ultra-flat, holey carbon-coated support grid for transmission electron microscopy (TEM). Unlike competing holey carbon films, C-flat is manufactured without plastics, so it is clean upon arrival and the user has no residue to contend with. Made with patent pending technology, C-flat provides an ultra-flat surface that results in better particle dispersion and more uniform ice thickness.
Patterning is done using deep-UV projection lithography, ensuring the most accurate and consistent hole shapes and sizes down to submicron features. The precise methods by which C-flat is manufactured elminate artifacts such as excess carbon and edges around holes.
C-flat holey carbon grids provide the ideal specimen support to achieve high resolution data in making them an ideal choice for cryo single particle analysis, cryo tomography and automated TEM analysis. 2.0 µm holes with 4.0 µm spacing.
Mesh: 400
Material: Copper
Catalog No. 50-192-6559 Supplier Electron Microscopy Sciences Supplier No. CFT44250
C-flat is an ultra-flat, holey carbon-coated support grid for transmission electron microscopy (TEM). Unlike competing holey carbon films, C-flat is manufactured without plastics, so it is clean upon arrival and the user has no residue to contend with. Made with patent pending technology, C-flat provides an ultra-flat surface that results in better particle dispersion and more uniform ice thickness.
Patterning is done using deep-UV projection lithography, ensuring the most accurate and consistent hole shapes and sizes down to submicron features. The precise methods by which C-flat is manufactured elminate artifacts such as excess carbon and edges around holes.
C-flat holey carbon grids provide the ideal specimen support to achieve high resolution data in making them an ideal choice for cryo single particle analysis, cryo tomography and automated TEM analysis. 2.0 µm holes with 4.0 µm spacing.
Mesh: 400
Material: Copper