Learn More
Accuflow Systems Saint-Gobain C-Flex Tubing, TPE, 3/8 in ID, 5/8 in OD
Catalog No. 502365933
Encompass_Preferred
C-Flex 374 TPE tubing is a pharmaceutical-grade thermoplastic elastomer tube formulated for bioprocess and laboratory fluid transfer. It is intended for aseptic welding and sealing disconnection in single-use assemblies, offering flexibility and reduced permeability compared with silicone.
  • Pharmaceutical-grade thermoplastic elastomer (TPE).
  • Translucent tubing, 0.375 in ID x 0.625 in OD.
  • Supplied as a 350 ft coil.
  • Sterilizable by gamma irradiation, X-ray irradiation, and autoclave.
  • Remains flexible from -45°C to 135°C (-50°F to 275°F).
  • Complies with USP Class VI and applicable ISO standards; animal-derived component-free.
  • Designed for aseptic welding, sealing disconnection, and single-use assemblies.
  • Lower permeability than silicone, suitable for high-purity fluid transfer.
Catalog No. 50-236-5933 Supplier Accuflow Systems Supplier No. 3743754S
C-Flex 374 TPE tubing is a pharmaceutical-grade thermoplastic elastomer tube formulated for bioprocess and laboratory fluid transfer. It is intended for aseptic welding and sealing disconnection in single-use assemblies, offering flexibility and reduced permeability compared with silicone.
  • Pharmaceutical-grade thermoplastic elastomer (TPE).
  • Translucent tubing, 0.375 in ID x 0.625 in OD.
  • Supplied as a 350 ft coil.
  • Sterilizable by gamma irradiation, X-ray irradiation, and autoclave.
  • Remains flexible from -45°C to 135°C (-50°F to 275°F).
  • Complies with USP Class VI and applicable ISO standards; animal-derived component-free.
  • Designed for aseptic welding, sealing disconnection, and single-use assemblies.
  • Lower permeability than silicone, suitable for high-purity fluid transfer.