Learn More
Accuflow Systems Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer1/4 in.idx7/16 in.od x 300 ft spool
Catalog No. 501048135
Encompass_Preferred
Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer1/4 in.idx7/16 in.od x 300 ft spool. Shore A 60 durometer,made with animal free compounds, ClassVI approved
Catalog No. 50-104-8135 Supplier Accuflow Systems Supplier No. 3742503M
Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer1/4 in.idx7/16 in.od x 300 ft spool. Shore A 60 durometer,made with animal free compounds, ClassVI approved