Learn More
Accuflow Systems Saint-Gobain C-Flex Tubing, TPE, 1/8 in ID, 1/4 in OD
Catalog No. NC3762510
Encompass_Preferred
C-Flex 082 TPE tubing is a biopharmaceutical-grade thermoplastic elastomer tubing supplied on 2,500 ft (762 m) spools for process and production fluid transfer. The 082 formulation provides chemical resistance and flexibility for use in pharmaceutical and bioprocess systems.
  • Biopharmaceutical-grade thermoplastic elastomer formulation 082.
  • Inner diameter 1/8 in (3.2 mm), outer diameter 1/4 in (6.4 mm), wall 0.063 in.
  • Supplied on 2,500 ft (762 m) spool for process-scale use.
  • Compatible with autoclave and gamma irradiation; suitable for single-use assemblies.
  • Supported by USP Class VI and ISO 10993 biocompatibility testing.
  • Operating temperature range approximately -45°C to 135°C (-50°F to 275°F).
Catalog No. NC3762510 Supplier Accuflow Systems Supplier No. 0821252S
C-Flex 082 TPE tubing is a biopharmaceutical-grade thermoplastic elastomer tubing supplied on 2,500 ft (762 m) spools for process and production fluid transfer. The 082 formulation provides chemical resistance and flexibility for use in pharmaceutical and bioprocess systems.
  • Biopharmaceutical-grade thermoplastic elastomer formulation 082.
  • Inner diameter 1/8 in (3.2 mm), outer diameter 1/4 in (6.4 mm), wall 0.063 in.
  • Supplied on 2,500 ft (762 m) spool for process-scale use.
  • Compatible with autoclave and gamma irradiation; suitable for single-use assemblies.
  • Supported by USP Class VI and ISO 10993 biocompatibility testing.
  • Operating temperature range approximately -45°C to 135°C (-50°F to 275°F).