Learn More
Accuflow Systems Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer3/8 in.idx5/8 in.od x 350 ft spool.
Catalog No. 501048140
Encompass_Preferred
Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer3/8 in.idx5/8 in.od x 350 ft spool. Shore A 60 durometer,made with animal free compounds, ClassVI approved
Catalog No. 50-104-8140 Supplier Accuflow Systems Supplier No. 0823754S
Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer3/8 in.idx5/8 in.od x 350 ft spool. Shore A 60 durometer,made with animal free compounds, ClassVI approved