Learn More
Accuflow Systems Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer1/8 in.idx1/4 in.od x 500 ft spool
Catalog No. 501048134
Encompass_Preferred
Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer1/8 in.idx1/4 in.od x 500 ft spool. Shore A 60 durometer,made with animal free compounds, ClassVI approved
Catalog No. 50-104-8134 Supplier Accuflow Systems Supplier No. 0821252M
Cole Parmer - CFLEX TUBING..Weldable and sealable Thermoplastic Elastomer1/8 in.idx1/4 in.od x 500 ft spool. Shore A 60 durometer,made with animal free compounds, ClassVI approved