Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B515 aHu CD11b Anti

Catalog No. 5000086928
Encompass_Preferred

Product Name: cFluor B515 Anti-Human CD11b (ICRF44)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Human CD11b (ICRF44)
Cytek Catalog Number: R7-20688
Marker: CD11b
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque|Pig
Clonality: Monoclonal
Clone: ICRF44
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B515
Size: 25 Tests

Catalog No. 50-000-86928 Supplier Cytek Biosciences Inc Supplier No. R720688
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor B515 Anti-Human CD11b (ICRF44)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Human CD11b (ICRF44)
Cytek Catalog Number: R7-20688
Marker: CD11b
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque|Pig
Clonality: Monoclonal
Clone: ICRF44
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B515
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.