Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B515 aHu CD5 Anti

Supplier:  Cytek Biosciences Inc R720142

Encompass_Preferred

Product Name: cFluor B515 Anti-Human CD5 (L17F12)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Human CD5 (L17F12)
Cytek Catalog Number: R7-20142
Marker: CD5
Target Species: Human
Clonality: Monoclonal
Clone: L17F12
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor B515
Size: 25 Tests

Catalog No. 50-000-86863


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.