Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B515 aHu IgM Anti

Catalog No. 5000086847
Encompass_Preferred

Product Name: cFluor B515 Anti-Human IgM (CH2)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Human IgM (CH2)
Cytek Catalog Number: R7-20287
Marker: IgM
Target Species: Human
Clonality: Monoclonal
Clone: CH2
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor B515
Size: 100 Tests

Catalog No. 50-000-86847 Supplier Cytek Biosciences Inc Supplier No. R720287
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor B515 Anti-Human IgM (CH2)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Human IgM (CH2)
Cytek Catalog Number: R7-20287
Marker: IgM
Target Species: Human
Clonality: Monoclonal
Clone: CH2
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor B515
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.