Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B515 aMu CD3 Anti

Supplier:  Cytek Biosciences Inc R720718

Encompass_Preferred

Product Name: cFluor B515 Anti-Mouse CD3 (17A2)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Mouse CD3 (17A2)
Cytek Catalog Number: R7-20718
Marker: CD3
Target Species: Mouse
Clonality: Monoclonal
Clone: 17A2
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor B515
Size: 25 Tests

Catalog No. 50-000-87132


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.