Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B515 aMu CD8a Anti

Catalog No. 5000087118
Encompass_Preferred

Product Name: cFluor B515 Anti-Mouse CD8a (53-6.7)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Mouse CD8a (53-6.7)
Cytek Catalog Number: R7-20545
Marker: CD8a
Target Species: Mouse
Clonality: Monoclonal
Clone: 53-6.7
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor B515
Size: 100 Tests

Catalog No. 50-000-87118 Supplier Cytek Biosciences Inc Supplier No. R720545
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor B515 Anti-Mouse CD8a (53-6.7)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Mouse CD8a (53-6.7)
Cytek Catalog Number: R7-20545
Marker: CD8a
Target Species: Mouse
Clonality: Monoclonal
Clone: 53-6.7
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor B515
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.