Learn More
Cytek Biosciences Inc cFluor B532 aHu CD14 Anti
Supplier: Cytek Biosciences Inc R720694
Product Name: cFluor B532 Anti-Human CD14 (MEM-18)
Product Class: Antibody
Type: Primary
Product Name: cFluor B532 Anti-Human CD14 (MEM-18)
Cytek Catalog Number: R7-20694
Marker: CD14
Target Species: Human
Clonality: Monoclonal
Clone: MEM-18
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor B532
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.