Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B532 aHu CD3 Anti

Catalog No. 5000087281
Encompass_Preferred

Product Name: cFluor B532 Anti-Human CD3 (UCHT1)
Product Class: Antibody
Type: Primary
Product Name: cFluor B532 Anti-Human CD3 (UCHT1)
Cytek Catalog Number: R7-21346
Marker: CD3
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: UCHT1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B532
Size: 25 Tests

Catalog No. 50-000-87281 Supplier Cytek Biosciences Inc Supplier No. R721346
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor B532 Anti-Human CD3 (UCHT1)
Product Class: Antibody
Type: Primary
Product Name: cFluor B532 Anti-Human CD3 (UCHT1)
Cytek Catalog Number: R7-21346
Marker: CD3
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: UCHT1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B532
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.