Learn More
Cytek Biosciences Inc cFluor B548 aHu CD45 Anti
Supplier: Cytek Biosciences Inc R720291
Product Name: cFluor B548 Anti-Human CD45 (HI30)
Product Class: Antibody
Type: Primary
Product Name: cFluor B548 Anti-Human CD45 (HI30)
Cytek Catalog Number: R7-20291
Marker: CD45
Target Species: Human
Clonality: Monoclonal
Clone: HI30
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B548
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.