Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B548 aHu CD45 Anti

Catalog No. 5000086828
Encompass_Preferred

Product Name: cFluor B548 Anti-Human CD45 (HI30)
Product Class: Antibody
Type: Primary
Product Name: cFluor B548 Anti-Human CD45 (HI30)
Cytek Catalog Number: R7-20292
Marker: CD45
Target Species: Human
Clonality: Monoclonal
Clone: HI30
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B548
Size: 25 Tests

Catalog No. 50-000-86828 Supplier Cytek Biosciences Inc Supplier No. R720292
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor B548 Anti-Human CD45 (HI30)
Product Class: Antibody
Type: Primary
Product Name: cFluor B548 Anti-Human CD45 (HI30)
Cytek Catalog Number: R7-20292
Marker: CD45
Target Species: Human
Clonality: Monoclonal
Clone: HI30
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B548
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.