Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B548 aMu Ly-6G Anti

Catalog No. 5000087295
Encompass_Preferred

Product Name: cFluor B548 Anti-Mouse Ly-6G (1A8)
Product Class: Antibody
Type: Primary
Product Name: cFluor B548 Anti-Mouse Ly-6G (1A8)
Cytek Catalog Number: R7-20544
Marker: Ly-6G
Target Species: Mouse
Clonality: Monoclonal
Clone: 1A8
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor B548
Size: 25 Tests

Catalog No. 50-000-87295 Supplier Cytek Biosciences Inc Supplier No. R720544
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor B548 Anti-Mouse Ly-6G (1A8)
Product Class: Antibody
Type: Primary
Product Name: cFluor B548 Anti-Mouse Ly-6G (1A8)
Cytek Catalog Number: R7-20544
Marker: Ly-6G
Target Species: Mouse
Clonality: Monoclonal
Clone: 1A8
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor B548
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.