Learn More
Cytek Biosciences Inc cFluor™ B548 Anti-Human CD4 SK3, 5 μL per test, 100 tests per vial, 59 kDa antigen, for flow cytometry
Catalog No. NC3234746
Encompass_Preferred
cFluor™ B548 Anti-Human CD4 SK3, 5 μL per test, 100 tests per vial, 59 kDa antigen, for flow cytometry
Catalog No. NC3234746 Supplier Cytek Biosciences Inc Supplier No. R720043
cFluor™ B548 Anti-Human CD4 SK3, 5 μL per test, 100 tests per vial, 59 kDa antigen, for flow cytometry