Learn More
Cytek Biosciences Inc cFluor B675 aHu CD4 Anti
Supplier: Cytek Biosciences Inc R720151
Product Name: cFluor B675 Anti-Human CD4 (SK3)
Product Class: Antibody
Type: Primary
Product Name: cFluor B675 Anti-Human CD4 (SK3)
Cytek Catalog Number: R7-20151
Marker: CD4
Target Species: Human
Clonality: Monoclonal
Clone: SK3
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B675
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.