Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B690 aHu Ig Lambda Anti

Catalog No. 5000086812
Encompass_Preferred

Product Name: cFluor B690 Anti-Human Ig Lambda (1-155-2)
Product Class: Antibody
Type: Primary
Product Name: cFluor B690 Anti-Human Ig Lambda (1-155-2)
Cytek Catalog Number: R7-20259
Marker: Ig Lambda
Target Species: Human
Clonality: Monoclonal
Clone: 1-155-2
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B690
Size: 100 Tests

Catalog No. 50-000-86812 Supplier Cytek Biosciences Inc Supplier No. R720259
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor B690 Anti-Human Ig Lambda (1-155-2)
Product Class: Antibody
Type: Primary
Product Name: cFluor B690 Anti-Human Ig Lambda (1-155-2)
Cytek Catalog Number: R7-20259
Marker: Ig Lambda
Target Species: Human
Clonality: Monoclonal
Clone: 1-155-2
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B690
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.