Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor B690 aHu Ig Lambda Anti

Supplier:  Cytek Biosciences Inc R720260

Encompass_Preferred

Product Name: cFluor B690 Anti-Human Ig Lambda (1-155-2)
Product Class: Antibody
Type: Primary
Product Name: cFluor B690 Anti-Human Ig Lambda (1-155-2)
Cytek Catalog Number: R7-20260
Marker: Ig Lambda
Target Species: Human
Clonality: Monoclonal
Clone: 1-155-2
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor B690
Size: 25 Tests

Catalog No. 50-000-87000


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.