Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG575 aHu CD84 Anti

Supplier:  Cytek Biosciences Inc R720459

Encompass_Preferred

Product Name: cFluor BYG575 Anti-Human CD84 (CD84.1.21)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG575 Anti-Human CD84 (CD84.1.21)
Cytek Catalog Number: R7-20459
Marker: CD84
Target Species: Human
Clonality: Monoclonal
Clone: CD84.1.21
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor BYG575
Size: 100 Tests

Catalog No. 50-000-86653


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.