Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG610 aHu CD27 Anti

Catalog No. 5000086902
Encompass_Preferred

Product Name: cFluor BYG610 Anti-Human CD27 (O323)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG610 Anti-Human CD27 (O323)
Cytek Catalog Number: R7-20399
Marker: CD27
Target Species: Human
Clonality: Monoclonal
Clone: O323
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor BYG610
Size: 100 Tests

Catalog No. 50-000-86902 Supplier Cytek Biosciences Inc Supplier No. R720399
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor BYG610 Anti-Human CD27 (O323)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG610 Anti-Human CD27 (O323)
Cytek Catalog Number: R7-20399
Marker: CD27
Target Species: Human
Clonality: Monoclonal
Clone: O323
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor BYG610
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.