Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG610 aHu CD33 Anti

Supplier:  Cytek Biosciences Inc R720022

Encompass_Preferred

Product Name: cFluor BYG610 Anti-Human CD33 (WM53)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG610 Anti-Human CD33 (WM53)
Cytek Catalog Number: R7-20022
Marker: CD33
Target Species: Human
Clonality: Monoclonal
Clone: WM53
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor BYG610
Size: 25 Tests

Catalog No. 50-000-86868


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.