Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG645 aMu TCR beta Ant

Catalog No. 5000087131
Encompass_Preferred

Product Name: cFluor BYG645 Anti-Mouse TCR beta (H57-597)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG645 Anti-Mouse TCR beta (H57-597)
Cytek Catalog Number: R7-20713
Marker: TCR beta
Target Species: Mouse
Clonality: Monoclonal
Clone: H57-597
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: cFluor BYG645
Size: 100 Tests

Catalog No. 50-000-87131 Supplier Cytek Biosciences Inc Supplier No. R720713
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor BYG645 Anti-Mouse TCR beta (H57-597)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG645 Anti-Mouse TCR beta (H57-597)
Cytek Catalog Number: R7-20713
Marker: TCR beta
Target Species: Mouse
Clonality: Monoclonal
Clone: H57-597
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: cFluor BYG645
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.