Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG667 aHu CD193 Anti

Catalog No. 5000087215
Encompass_Preferred

Product Name: cFluor BYG667 Anti-Human CD193 (CCR3) (5E8)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG667 Anti-Human CD193 (CCR3) (5E8)
Cytek Catalog Number: R7-20455
Marker: CD193 (CCR3)
Target Species: Human
Clonality: Monoclonal
Clone: 5E8
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG667
Size: 100 Tests

Catalog No. 50-000-87215 Supplier Cytek Biosciences Inc Supplier No. R720455
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor BYG667 Anti-Human CD193 (CCR3) (5E8)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG667 Anti-Human CD193 (CCR3) (5E8)
Cytek Catalog Number: R7-20455
Marker: CD193 (CCR3)
Target Species: Human
Clonality: Monoclonal
Clone: 5E8
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG667
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.