Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG667 aMu CD317 Anti

Catalog No. 5000086508
Encompass_Preferred

Product Name: cFluor BYG667 Anti-Mouse CD317 (927)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG667 Anti-Mouse CD317 (927)
Cytek Catalog Number: R7-20554
Marker: CD317
Target Species: Mouse
Clonality: Monoclonal
Clone: 927
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor BYG667
Size: 25 Tests

Catalog No. 50-000-86508 Supplier Cytek Biosciences Inc Supplier No. R720554
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor BYG667 Anti-Mouse CD317 (927)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG667 Anti-Mouse CD317 (927)
Cytek Catalog Number: R7-20554
Marker: CD317
Target Species: Mouse
Clonality: Monoclonal
Clone: 927
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor BYG667
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.