Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG710 aHu CD181 Anti

Catalog No. 5000087197
Encompass_Preferred

Product Name: cFluor BYG710 Anti-Human CD181 (CXCR1) (8F1-1-4)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG710 Anti-Human CD181 (CXCR1) (8F1-1-4)
Cytek Catalog Number: R7-20338
Marker: CD181 (CXCR1)
Target Species: Human
Clonality: Monoclonal
Clone: 8F1-1-4
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor BYG710
Size: 25 Tests

Catalog No. 50-000-87197 Supplier Cytek Biosciences Inc Supplier No. R720338
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor BYG710 Anti-Human CD181 (CXCR1) (8F1-1-4)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG710 Anti-Human CD181 (CXCR1) (8F1-1-4)
Cytek Catalog Number: R7-20338
Marker: CD181 (CXCR1)
Target Species: Human
Clonality: Monoclonal
Clone: 8F1-1-4
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor BYG710
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.