Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG710 aHu CD25 Anti

Supplier:  Cytek Biosciences Inc R720660

Encompass_Preferred

Product Name: cFluor BYG710 Anti-Human CD25 (4E3)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG710 Anti-Human CD25 (4E3)
Cytek Catalog Number: R7-20660
Marker: CD25
Target Species: Human
Reported Cross Reactivity: Rhesus Macaque
Clonality: Monoclonal
Clone: 4E3
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor BYG710
Size: 25 Tests

Catalog No. 50-000-87314


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.