Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG710 aMu CD49b Anti

Catalog No. 5000086654
Encompass_Preferred

Product Name: cFluor BYG710 Anti-Mouse CD49b (HMa2)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG710 Anti-Mouse CD49b (HMa2)
Cytek Catalog Number: R7-20577
Marker: CD49b
Target Species: Mouse
Clonality: Monoclonal
Clone: HMa2
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: cFluor BYG710
Size: 100 Tests

Catalog No. 50-000-86654 Supplier Cytek Biosciences Inc Supplier No. R720577
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor BYG710 Anti-Mouse CD49b (HMa2)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG710 Anti-Mouse CD49b (HMa2)
Cytek Catalog Number: R7-20577
Marker: CD49b
Target Species: Mouse
Clonality: Monoclonal
Clone: HMa2
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: cFluor BYG710
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.