Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG750 aHu CD13 Anti

Supplier:  Cytek Biosciences Inc R720733

Encompass_Preferred

Product Name: cFluor BYG750 Anti-Human CD13 (WM15)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Human CD13 (WM15)
Cytek Catalog Number: R7-20733
Marker: CD13
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: WM15
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG750
Size: 100 Tests

Catalog No. 50-000-87053


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.