Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG750 aHu CD195 Anti

Catalog No. 5000087117
Encompass_Preferred

Product Name: cFluor BYG750 Anti-Human CD195 (CCR5) (HEK/1/85a)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Human CD195 (CCR5) (HEK/1/85a)
Cytek Catalog Number: R7-20481
Marker: CD195 (CCR5)
Target Species: Human
Clonality: Monoclonal
Clone: HEK/1/85a
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor BYG750
Size: 100 Tests

Catalog No. 50-000-87117 Supplier Cytek Biosciences Inc Supplier No. R720481
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor BYG750 Anti-Human CD195 (CCR5) (HEK/1/85a)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Human CD195 (CCR5) (HEK/1/85a)
Cytek Catalog Number: R7-20481
Marker: CD195 (CCR5)
Target Species: Human
Clonality: Monoclonal
Clone: HEK/1/85a
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor BYG750
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.