Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG750 aHu CD300e Anti

Catalog No. 5000087313
Encompass_Preferred

Product Name: cFluor BYG750 Anti-Human CD300e (UP-H2)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Human CD300e (UP-H2)
Cytek Catalog Number: R7-20591
Marker: CD300e
Target Species: Human
Clonality: Monoclonal
Clone: UP-H2
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG750
Size: 100 Tests

Catalog No. 50-000-87313 Supplier Cytek Biosciences Inc Supplier No. R720591
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor BYG750 Anti-Human CD300e (UP-H2)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Human CD300e (UP-H2)
Cytek Catalog Number: R7-20591
Marker: CD300e
Target Species: Human
Clonality: Monoclonal
Clone: UP-H2
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG750
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.