Learn More
Cytek Biosciences Inc cFluor BYG750 aHu IgD Anti
Supplier: Cytek Biosciences Inc R720662
Product Name: cFluor BYG750 Anti-Human IgD (IgD26)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Human IgD (IgD26)
Cytek Catalog Number: R7-20662
Marker: IgD
Target Species: Human
Clonality: Monoclonal
Clone: IgD26
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG750
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.