Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG750 aMu MHCII Anti

Catalog No. 5000087156
Encompass_Preferred

Product Name: cFluor BYG750 Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Cytek Catalog Number: R7-20564
Marker: MHC Class II (I-A/I-E)
Target Species: Mouse
Clonality: Monoclonal
Clone: M5/114.15.2
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor BYG750
Size: 25 Tests

Catalog No. 50-000-87156 Supplier Cytek Biosciences Inc Supplier No. R720564
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor BYG750 Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Cytek Catalog Number: R7-20564
Marker: MHC Class II (I-A/I-E)
Target Species: Mouse
Clonality: Monoclonal
Clone: M5/114.15.2
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor BYG750
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.