Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG781 aHu CD14 Anti

Catalog No. 5000087081
Encompass_Preferred

Product Name: cFluor BYG781 Anti-Human CD14 (MEM-15)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG781 Anti-Human CD14 (MEM-15)
Cytek Catalog Number: R7-20325
Marker: CD14
Target Species: Human
Clonality: Monoclonal
Clone: MEM-15
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG781
Size: 100 Tests

Catalog No. 50-000-87081 Supplier Cytek Biosciences Inc Supplier No. R720325
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor BYG781 Anti-Human CD14 (MEM-15)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG781 Anti-Human CD14 (MEM-15)
Cytek Catalog Number: R7-20325
Marker: CD14
Target Species: Human
Clonality: Monoclonal
Clone: MEM-15
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor BYG781
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.