Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor BYG781 aMu CD62L Anti

Catalog No. 5000087222
Encompass_Preferred

Product Name: cFluor BYG781 Anti-Mouse CD62L (L-Selectin) (MEL-14)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG781 Anti-Mouse CD62L (L-Selectin) (MEL-14)
Cytek Catalog Number: R7-20565
Marker: CD62L (L-Selectin)
Target Species: Mouse
Clonality: Monoclonal
Clone: MEL-14
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor BYG781
Size: 100 Tests

Catalog No. 50-000-87222 Supplier Cytek Biosciences Inc Supplier No. R720565
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor BYG781 Anti-Mouse CD62L (L-Selectin) (MEL-14)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG781 Anti-Mouse CD62L (L-Selectin) (MEL-14)
Cytek Catalog Number: R7-20565
Marker: CD62L (L-Selectin)
Target Species: Mouse
Clonality: Monoclonal
Clone: MEL-14
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor BYG781
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.