Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R659 aHu CD34 Anti

Supplier:  Cytek Biosciences Inc R720676

Encompass_Preferred

Product Name: cFluor R659 Anti-Human CD34 (4H11)
Product Class: Antibody
Type: Primary
Product Name: cFluor R659 Anti-Human CD34 (4H11)
Cytek Catalog Number: R7-20676
Marker: CD34
Target Species: Human
Clonality: Monoclonal
Clone: 4H11
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R659
Size: 25 Tests

Catalog No. 50-000-87240

Please call customer service for current pricing. 1-800-766-7000


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.